Phthalimide DBU salt ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Phthalimide 1,8-Diazabicyclo[5.4.0]undec-7-ene Salt, DBU Phthalimide Salt

P1235-25G
TCIAP1235-25GEA 1175 NOK
TCIAP1235-25G
Phthalimide DBU salt ≥98.0% (by titrimetric analysis)
Phthalimide DBU salt
Formel: C₁₇H₂₁N₃O₂
MW: 299,37 g/mol
MDL nummer: MFCD00191683
CAS-nummer: 119812-51-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR