m,m,m-Tritolyl phosphate ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: m,m,m-TCP, Phosphoric acid tri-m-tolyl ester, Tris(3-methylphenyl) phosphate, Phosphoric acid tri-m-cresyl ester, Tri-m-tolyl phosphate, Tri-m-tolylphosphate, m,m,m-Tricresyl phosphate, Tri-m-cresyl phosphate

P1472-5G
TCIAP1472-5GEA 1500 NOK
TCIAP1472-5G
m,m,m-Tritolyl phosphate ≥95.0% (by GC)
m,m,m-Tritolyl phosphate
Formel: C₂₁H₂₁O₄P
Kokepunkt: 240°C
Smeltepunkt: 25…26 °C
Tetthet: 1,15 g/cm³ (25 °C)
MDL nummer: MFCD00041907
CAS-nummer: 563-04-2
EINECS: 209-241-8
FN: 3077
ADR: 9,III
Merck Indeks: 14,09763
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95292e69366473d76f26b46e7a650650c0cbc0d0&vid=95292e69366473d76f26b46e7a650650c0cbc0d0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to lump
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR