Pyridine hydrobromide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

P1666-25G
TCIAP1666-25GEA 220 NOK
TCIAP1666-25G
Pyridine hydrobromide ≥98.0% (by HPLC, titration analysis)
Pyridine hydrobromide
Formel: C₅H₅N
Smeltepunkt: 217°C
MDL nummer: MFCD00012801
CAS-nummer: 18820-82-1
EINECS: 242-600-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 216.0 to 221.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR