2-Phenyl-5-benzimidazolesulphonic acid ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Ensulizole, Phenylbenzimidazole sulphonic acid, 1H-Benzimidazole-5-sulphonic acid

P1670-25G
TCIAP1670-25GEA 395 NOK
TCIAP1670-25G
2-Phenyl-5-benzimidazolesulphonic acid ≥95.0% (by titrimetric analysis)
2-Phenyl-5-benzimidazolesulphonic acid
Formel: C₁₃H₁₀N₂O₃S
MW: 274,29 g/mol
Smeltepunkt: 410°C
MDL nummer: MFCD00053007
CAS-nummer: 27503-81-7
EINECS: 248-502-0
Merck Indeks: 14,03593
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=296bf70667798de8a89db49f76c9fc12df1818aa&vid=da96b7aacb76433fe9d64b6a04a3c540530ff085&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 94.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR