Penicillin G kaliumsalt ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Danger

Synonymer: 6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt

P1772-25G
TCIAP1772-25GEA 515 NOK
TCIAP1772-25G
Penicillin G kaliumsalt ≥98.0% (by HPLC, total nitrogen)
Penicillin G kaliumsalt
Formel: C₁₆H₁₇KN₂O₄S
Smeltepunkt: 217°C
Tetthet: 1,359 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036193
CAS-nummer: 113-98-4
EINECS: 204-038-0
Merck Indeks: 14,07094
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83141b7587d6e0c38f8171e1c85e0961556dfe90&vid=83141b7587d6e0c38f8171e1c85e0961556dfe90&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +270.0 to +300.0 deg (C=1, H2O)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR