1,10-Fenatrolin monohydrat ≥99.0% (by titrimetric analysis) for biochemical research

Leverandør: TCI
Danger

Synonymer: 1,10-Phenanthroline hydrate

P1826-5G
TCIAP1826-5GEA 310 NOK
TCIAP1826-5G
1,10-Fenatrolin monohydrat ≥99.0% (by titrimetric analysis) for biochemical research
1,10-Fenatrolin monohydrat
Formel: C₁₂H₈N₂·1H₂O
MW: 198,22 g/mol
Smeltepunkt: 102°C
Tetthet: 1,259 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149973
CAS-nummer: 5144-89-8
EINECS: 200-629-2
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,07214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=885a0f0fae4ef9fd25401df9e424ed9b90c01f94&vid=1e4e13c368085e4d5577a8d867ac5ca0920fdeeb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Enzyme inhibition test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR