2-(1-Piperazinylcarbonyl)-1,4-benzodioxane ≥95.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine

P1836-5G
TCIAP1836-5GEA 1200 NOK
TCIAP1836-5G
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane ≥95.0% (by GC, titration analysis)
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane
Formel: C₁₃H₁₆N₂O₃
MW: 248,28 g/mol
MDL nummer: MFCD01837119
CAS-nummer: 70918-00-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR