4-(Phenylsulphonyl)-2-azetidinone ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Danger

P1895-1G
TCIAP1895-1GEA 985 NOK
TCIAP1895-1G
4-(Phenylsulphonyl)-2-azetidinone ≥98.0% (by HPLC, total nitrogen)
4-(Phenylsulphonyl)-2-azetidinone
Formel: C₉H₉NO₃S
MW: 211,24 g/mol
MDL nummer: MFCD00272351
CAS-nummer: 31899-01-1
FN: 1759
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 153.0 to 157.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR