Phenylsulphur pentafluoride ≥95.0% (by GC)

Leverandør: TCI
Warning

P1969-1G
TCIAP1969-1GEA 1225 NOK
TCIAP1969-1G
Phenylsulphur pentafluoride ≥95.0% (by GC)
Phenylsulphur pentafluoride
Formel: C₆H₅F₅S
Kokepunkt: 149°C
Smeltepunkt: –10°C
Flash punkt: 48°C
MDL nummer: MFCD09263740
CAS-nummer: 2557-81-5
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4873d97bc60eea87475d31806440714d279fd6d8&vid=4873d97bc60eea87475d31806440714d279fd6d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR