4-Pentylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

P2045-5G
TCIAP2045-5GEA 2800 NOK
TCIAP2045-5G
4-Pentylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-Pentylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₂H₂₂O₂
MDL nummer: MFCD00466238
CAS-nummer: 38792-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8178fbdb1d2e7a57135e64d75e24f1d73dd41df4&vid=8178fbdb1d2e7a57135e64d75e24f1d73dd41df4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR