3,4,9,10-Perylenetetracarboxylic dianhydride ≥98.0% (by HPLC), purified by sublimation

Leverandør: TCI
Warning

Synonymer: Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA

P2102-1G
TCIAP2102-1GEA 1125 NOK
TCIAP2102-1G
3,4,9,10-Perylenetetracarboxylic dianhydride ≥98.0% (by HPLC), purified by sublimation
3,4,9,10-Perylenetetracarboxylic dianhydride
Formel: C₂₄H₈O₆
MW: 392,32 g/mol
MDL nummer: MFCD00006916
CAS-nummer: 128-69-8
EINECS: 204-905-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26c7eeb906805ba1218fff3f0bb7fdb32cc58923&vid=26c7eeb906805ba1218fff3f0bb7fdb32cc58923&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Amber to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR