1,2-Diphenyl-1-butanone 98,0 GC_ASSAY_METHOD

Leverandør: TCI

Synonymer: 1,2-Diphenyl-1-butanone, 2-Phenylbutyrophenone, 1,2-Diphenylbutan-1-one

P2269-5G
TCIAP2269-5GEA 1000 NOK
TCIAP2269-5G
1,2-Diphenyl-1-butanone 98,0 GC_ASSAY_METHOD
1,2-Diphenyl-1-butanone
Formel: C₁₆H₁₆O
MW: 224.3000 g/mol
Kokepunkt: 324°C
MDL nummer: MFCD00010344
CAS-nummer: 16282-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f5e5beebc7e0687d53caf6b384760864df185ed&vid=9f5e5beebc7e0687d53caf6b384760864df185ed&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR