Poly(2-ethyl-2-oxazoline) n=∼100, ULTROXA®

Leverandør: TCI

P2507-500MG
TCIAP2507-500MGEA 2550 NOK
TCIAP2507-500MG
Poly(2-ethyl-2-oxazoline) n=∼100, ULTROXA®
Poly(2-ethyl-2-oxazoline)
Formel: (C₅H₉NO)x
MW: 200000 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 161358-47-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Average molecular weight 8,000 to 12,000
Molecular weight distribution max. 1.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR