2-Oleoyl-1-palmitoyl-Sn-glycero-3-phospho-rac-(1-glycerol)sodium salt 98,0 Titrimetric

Leverandør: TCI

Synonymer: POPG, 1-Palmitoyl-2-oleyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt, POPG sodium salt, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt), POPG-Na, 1-Palmitoyl-2-oleoyl phosphatidylglycerol sodium salt, Sodium 2,3-dihydroxypropyl ((R)-2-(oleoyloxy)-3-(palmitoyloxy)propyl) phosphate(Z), Sodium 2,3-Dihydroxypropyl [2-(Oleoyloxy)-3-(palmitoyloxy)propyl] phosphate

P2522-250MG
TCIAP2522-250MGEA 2925 NOK
TCIAP2522-250MG
2-Oleoyl-1-palmitoyl-Sn-glycero-3-phospho-rac-(1-glycerol)sodium salt 98,0 Titrimetric
2-Oleoyl-1-palmitoyl-Sn-glycero-3-phospho-rac-(1-glycerol)sodium salt
Formel: C₄₀H₇₆NaO₁₀P
MW: 771.002 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00674290
CAS-nummer: 268550-95-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=992644ea5a2fe3895ba40ce980e90e4954266e93&vid=992644ea5a2fe3895ba40ce980e90e4954266e93&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 94.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR