Poly(tert-Butyl Acrylate)-block-Poly(styrene) (Copolymer, 10:6)

Leverandør: TCI

P2553-200MG
TCIAP2553-200MGEA 1300 NOK
TCIAP2553-200MG
Poly(tert-Butyl Acrylate)-block-Poly(styrene) (Copolymer, 10:6)
Poly(tert-Butyl Acrylate)-block-Poly(styrene)
CAS-nummer: 697765-49-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Molecular weight of number average 15,000 to 20,000
Molecular weight distribution 1.00 to 1.40

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR