Pifithrin-α hydrobromide 98,0 HPLC, T

Leverandør: TCI

Synonymer: Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide

P2751-10MG P2751-50MG
TCIAP2751-10MGEA 1450 NOK
TCIAP2751-10MG TCIAP2751-50MG
Pifithrin-α hydrobromide 98,0 HPLC, T
Pifithrin-alpha hydrobromide
Formel: C₁₆H₁₈N₂OS·HBr
MW: 286.4·80.9 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00417851
CAS-nummer: 63208-82-2
Merck Indeks: 14,07421

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR