Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)

Leverandør: TCI
Warning

Q0023-25G
TCIAQ0023-25GEA 875 NOK
TCIAQ0023-25G
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)
Formel: C₁₈H₁₁NO₂.xNaO₃S
MDL nummer: MFCD00080727
CAS-nummer: 8004-92-0
Merck Indeks: 14,08071

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Lambda max. 410.0 to 414.0 nm (H2O)
Absorbance (E1%1cm) min. 600 (H2O, 410.0 to 414.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR