Quinoline-N-oxide hydrate ≥97.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Quinoline 1-oxide hydrate

Q0072-25G Q0072-5G
TCIAQ0072-25GEA 5875 NOK
TCIAQ0072-25G TCIAQ0072-5G
Quinoline-N-oxide hydrate ≥97.0% (by titrimetric analysis)
Quinoline-N-oxide hydrate
Formel: C₉H₇NO·xH₂O
Kokepunkt: 173°C
MDL nummer: MFCD00149472
CAS-nummer: 198878-42-1
EINECS: 216-560-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Brown powder to crystal
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Water 15.0 to 21.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR