Quetiapine hemifumarate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: ICI-204636, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol bis[(2E)-2-butenedioate], Seroquel

Q0092-5G Q0092-25G
TCIAQ0092-5GEA 1025 NOK
TCIAQ0092-5G TCIAQ0092-25G
Quetiapine hemifumarate ≥98.0% (by HPLC, titration analysis)
Quetiapine hemifumarate
Formel: C₂₁H₂₅N₃O₂S
MW: 441.55 g/mol
MDL nummer: MFCD03423782
CAS-nummer: 111974-72-2
Merck Indeks: 14,08039
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5865a5f1c59c4dba63f10a4f981a800fa9f1601b&vid=5865a5f1c59c4dba63f10a4f981a800fa9f1601b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR