Quinoxyfen ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (5,7-Dichloro-4-quinolyl) (4-Fluorophenyl) Ether, 5,7-Dichloro-4-(4-fluorophenoxy)quinoline

Q0093-200MG
TCIAQ0093-200MGEA 1000 NOK
TCIAQ0093-200MG
Quinoxyfen ≥98.0% (by GC)
Quinoxyfen
Formel: C₁₅H₈Cl₂FNO
MW: 308,13 g/mol
MDL nummer: MFCD03265638
CAS-nummer: 124495-18-7
FN: 3077
ADR: 9,III
Merck Indeks: 14,08079
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65c4e1a6ac58002963bfa476299e0979e3ebac9f&vid=65c4e1a6ac58002963bfa476299e0979e3ebac9f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR