Rhodamin B ≥95.0% (by HPLC)

Leverandør: TCI
Danger

Synonymer: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

R0040-25G R0040-250G
TCIAR0040-25GEA 405 NOK
TCIAR0040-25G TCIAR0040-250G
Rhodamin B ≥95.0% (by HPLC)
Rhodamin B
Formel: C₂₈H₃₁ClN₂O₃
Smeltepunkt: 199…201 °C
Tetthet: 1,31 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011931
CAS-nummer: 81-88-9
EINECS: 201-383-9
Merck Indeks: 14,08183
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daa0764afa6a24891ef507201d36103bd6e3c98d&vid=daa0764afa6a24891ef507201d36103bd6e3c98d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Lambda max. 552.0 to 556.0 nm (H2O)
Absorbance (E1%1cm) min. 2000 (H2O, 552.0 to 556.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR