Ruthenium(II) nitrosyl chloride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Trichloronitrosylruthenium(II), Ruthenium(II) nitrosylchloride

R0086-1G
TCIAR0086-1GEA 3825 NOK
TCIAR0086-1G
Ruthenium(II) nitrosyl chloride ≥97.0% (by titrimetric analysis)
Ruthenium(II) nitrosyl chloride
Formel: Ru(NO)Cl₃
MW: 237,43 g/mol
MDL nummer: MFCD00049765
CAS-nummer: 18902-42-6
EINECS: 242-651-5

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to Amber to Dark red powder to crystal
Purity (Argentmetric Titration) min. 97.0 % (calcd.on anh.substance)
Water 6.0 to 9.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR