Rhapontigenin ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (E)-5-(3-Hydroxy-4-methoxystyryl)benzene-1,3-diol, 3,3',5-Trihydroxy-4'-methoxy-trans-stilbene, trans-1-(3,5-Dihydroxyphenyl)-2-(3-hydroxy-4-methoxyphenyl)ethylene

R0089-100MG
TCIAR0089-100MGEA 2725 NOK
TCIAR0089-100MG
Rhapontigenin ≥98.0% (by GC)
Rhapontigenin
Formel: C₁₅H₁₄O₄
MW: 258,27 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00017718
CAS-nummer: 500-65-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR