Rapamycin ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone

R0097-25MG
TCIAR0097-25MGEA 945 NOK
TCIAR0097-25MG
Rapamycin ≥98.0% (by HPLC)
Rapamycin
Formel: C₅₁H₇₉NO₁₃
MW: 914.19 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00867594
CAS-nummer: 53123-88-9
Merck Indeks: 14,08114

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 1.20 to 1.70 %
Specific rotation [a]20/D -145 to -155 deg (C=0.2, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR