Rosuvastatin calcium salt ≥95.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: 7-{4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulphonyl)amino]pyrimidin-5-yl}-3,5-dihydroxyhept-6-enoic acid, Calcium (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulphonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate, Rosuvastatin Calcium, CRESTOR, (3R,5S,6E)-7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[N-methyl(n-methylsulphonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-Heptenoic calcium

R0180-200MG
TCIAR0180-200MGEA 1100 NOK
TCIAR0180-200MG
Rosuvastatin calcium salt ≥95.0% (by HPLC, total nitrogen)
Rosuvastatin calcium salt
Formel: C₂₂H₂₇FN₃O₆S·¹/₂Ca
Smeltepunkt: 160°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD04112702
CAS-nummer: 147098-20-2
Merck Indeks: 14,08270
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd6387b471437759fd7360cc3312748493df2f20&vid=a0faf79fc3b760f080680a89ff5bb7a36bc22d2d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Purity (Chelometric Titration) min. 90.0 %
Specific rotation [a]20/D -7.0 to -9.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR