Rhodamine 19 ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

R0186-1G R0186-200MG
TCIAR0186-1GEA 3250 NOK
TCIAR0186-1G TCIAR0186-200MG
Rhodamine 19 ≥97.0% (by HPLC, total nitrogen)
Rhodamine 19
Formel: C₂₆H₂₆N₂O₃
Smeltepunkt: 299°C
MDL nummer: MFCD09056722
CAS-nummer: 25152-49-2
EINECS: 246-666-8

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Molar extinction coefficient min. 13000 (MeOH, 516.0 to 521.0nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR