Rhodamine 640 Perchlorate ≥90.0% (by HPLC)

Leverandør: TCI
Warning

R0188-200MG R0188-1G
TCIAR0188-200MGEA 835 NOK
TCIAR0188-200MG TCIAR0188-1G
Rhodamine 640 Perchlorate ≥90.0% (by HPLC)
Rhodamine 640 Perchlorate
Formel: C₃₂H₃₁ClN₂O₇
MDL nummer: MFCD00799440
CAS-nummer: 72102-91-1
EINECS: 276-345-8
FN: 1479
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dac96f5894a02d9e9af53d44514f41752390a76&vid=7dac96f5894a02d9e9af53d44514f41752390a76&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Amber to Brown to Dark green powder to crystal
Purity (HPLC) min. 90.0 area%
Lambda max. 560.0 to 575.0 nm (MeOH)
Absorbance (E1%1cm) min. 1800 (MeOH, 560 to 575nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR