Rhodamine 700 90,0 HPLC_ASSAY_METHOD

Leverandør: TCI
Warning

R0189-200MG
TCIAR0189-200MGEA 2250 NOK
TCIAR0189-200MG
Rhodamine 700 90,0 HPLC_ASSAY_METHOD
Rhodamine 700
Formel: C₂₆H₂₆ClF₃N₂O₅
MW: 538.95 g/mol
Smeltepunkt: 205°C
CAS-nummer: 63561-42-2
EINECS: 264-312-0
FN: 1479
ADR: 5.1 III

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 90.0 area%
Lambda max. 643.0 to 651.0 nm (ethanol)
Molar extinction coefficient min. 90000 (ethanol, 643.0 ~ 651.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR