Rhodamine 19 perchlorate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

R0191-1G R0191-5G
TCIAR0191-1GEA 930 NOK
TCIAR0191-1G TCIAR0191-5G
Rhodamine 19 perchlorate ≥98.0% (by HPLC, total nitrogen)
Rhodamine 19 perchlorate
Formel: C₂₆H₂₇ClN₂O₇
MDL nummer: MFCD02684062
CAS-nummer: 62669-66-3
EINECS: 263-686-2
FN: 1479
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=015e6a32da526689073d1dd488bd23525a62aeae&vid=015e6a32da526689073d1dd488bd23525a62aeae&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 515.0 to 523.0 nm (MeOH)
Molar extinction coefficient min. 97000 (MeOH, 515.0 to 523.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR