Resazurin sodium salt ≥85.0% (by HPLC)

Leverandør: TCI
Warning

R0203-1G R0203-5G
TCIAR0203-1GEA 380 NOK
TCIAR0203-1G TCIAR0203-5G
Resazurin sodium salt ≥85.0% (by HPLC)
Resazurin sodium salt
Formel: C₁₂H₆NNaO₄
MDL nummer: MFCD00005036
CAS-nummer: 62758-13-8
EINECS: 263-718-5
Merck Indeks: 14,08141
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84a9e1a6901d7e94e86443ffb641c17c336f8cd4&vid=84a9e1a6901d7e94e86443ffb641c17c336f8cd4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Dark green to Dark blue to Black powder to crystal
Purity (HPLC) min. 85.0 area%
Lambda max. 598.0 to 602.0nm (H2O)
Absorbance min. 0.450 (5mg/L、H2O、598.0 to 602.0 nm) (calcd.on dried substance)
Drying loss max. 10.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR