rPSL1a-Biotin Substrate for Sia alpha(2-6)Gal

Leverandør: TCI

Synonymer: rPSL1a-Biotin Conjugate, Recombinant Polyporus squamosus lectin (= rPSL1a)-Biotin Conjugate

R0230-1ML
TCIAR0230-1MLEA 4275 NOK
TCIAR0230-1ML
rPSL1a-Biotin Substrate for Sia alpha(2-6)Gal
rPSL1a-Biotin

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Lowry Methoed) 0.9 to 1.5 mg/mL
Suitability for use in ELISA tests min. 1.0 (1.0 micro g/mL, OD450)
Molecular weight 25 to 40 kDa


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR