rLSL-N-Biotin Substr.Gal beta(1-4)GlcNAc, poly LacNAc

Leverandør: TCI

Synonymer: Recombinant Laetiporus sulphureus lectin N-Terminal Domain (= rLSL-N)-Biotin Conjugate, rLSL-N-Biotin Conjugate

R0231-1ML
TCIAR0231-1MLEA 3300 NOK
TCIAR0231-1ML
rLSL-N-Biotin Substr.Gal beta(1-4)GlcNAc, poly LacNAc
rLSL-N-Biotin

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Lowry Methoed) 0.9 to 1.5 mg/mL
Molecular weight 10 to 25 kDa
Suitability for use in ELISA tests min. 1.0 (10 micro g/mL, OD450)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR