rMOA-Biotin for Gal alpha(1-3)Gal

Leverandør: TCI

R0232-1ML
TCIAR0232-1MLEA 3300 NOK
TCIAR0232-1ML
rMOA-Biotin for Gal alpha(1-3)Gal
rMOA-Biotin

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Lowry Methoed) 0.9 to 1.5 mg/mL
Molecular weight 25 to 40 kDa
Suitability for use in ELISA tests min. 1.0 (1.0 micro g/mL, OD450)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR