rSRL-Biotin Subst.GlcNAc B(1-2)Man, Gal B(1-3)GalNAc

Leverandør: TCI

Synonymer: rSRL-Biotin Conjugate, Recombinant Sclerotium rolfsii lectin (= rSRL)-Biotin Conjugate

R0233-1ML
TCIAR0233-1MLEA 3300 NOK
TCIAR0233-1ML
rSRL-Biotin Subst.GlcNAc B(1-2)Man, Gal B(1-3)GalNAc
rSRL-Biotin

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Lowry Methoed) 0.9 to 1.5 mg/mL
Molecular weight 10 to 25 kDa
Suitability for use in ELISA tests min. 1.0 (10 micro g/mL, OD450)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR