rGRFT-Biotin Substrate for Man alpha(1-2)Man

Leverandør: TCI

Synonymer: rGRFT-Biotin Conjugate, Recombinant Griffithsia sp. lectin (= rGRFT)-Biotin Conjugate, Recombinant Griffithsin-Biotin Conjugate

R0234-1ML
TCIAR0234-1MLEA 3300 NOK
TCIAR0234-1ML
rGRFT-Biotin Substrate for Man alpha(1-2)Man
rGRFT-Biotin

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Lowry Methoed) 0.9 to 1.5 mg/mL
Molecular weight 10 to 25 kDa
Suitability for use in ELISA tests min. 1.0 (1.0 micro g/mL, OD450)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR