rLSL-N-LecBeads [for Galβ(1-4)GlcNAc, poly LacNAc]

Leverandør: TCI

R0236-1VIAL
TCIAR0236-1VIALEA 4250 NOK
TCIAR0236-1VIAL
rLSL-N-LecBeads [for Galβ(1-4)GlcNAc, poly LacNAc]
Proteinrense Protein Purification Resins

The rLSL-N is an Escherichia coli recombinant of N-terminal domain of carbohydrate-binding protein (Lectin) derived from Laetiporus sulphureus. This lectin preferably binds to [Galβ(1-4)GlcNAc, poly LacNAc] glycan epitopes (It very slightly interacts with lactose).
The LecBeads is a lectin-immobilized agarose which is capable of capturing glycoproteins via binding to its specific epitope(s), and it is prepared by a covalent coupling method between lectin and agarose.
The Lectin-catch method using LecBeads (as shown in Application tab) is available for selective separation of glycoconjugates (glycoproteins, glycolipids, and so on). This LSL-N-LecBeads (1 ml gel) shows a binding capacity of about 12 mg or more asialo human Transferrin [asialo hTF] through its binding to a terminal [Galβ(1-4)GlcNAc] glycan epitope on asialo hTF.


  • Preparation : Recombinant LSL-N is covalently coupled with agarose gel
  • Quantity : 1 ml gel/2 ml (1:1, v/v slurry) suspended in PBS, pH7.5
  • Protein concentration : about 10 mg/ml gel
  • Binding capacity : >12 mg of glycoprotein (asialo human Transferrin) /ml gel
  • Preservative : 0.02% [w/v] Sodium azide
  • Stabilizer : None
  • Store at 2-10 °C. (Avoid freezing).

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR