rMOA-LecBeads [for Galα(1-3)Gal]

Leverandør: TCI

R0237-1VIAL
TCIAR0237-1VIALEA 4250 NOK
TCIAR0237-1VIAL
rMOA-LecBeads [for Galα(1-3)Gal]
Proteinrense Protein Purification Resins

The rMOA is an Escherichia coli recombinant of carbohydrate-binding protein (Lectin) derived from Marasmius oreades. This lectin preferably binds to a [Galα(1-3)Gal] glycan epitope (It very slightly interacts with melibiose).
The LecBeads is a lectin-immobilized agarose which is capable of capturing glycoproteins via binding to its specific epitope(s), and it is prepared by a covalent coupling method between lectin and agarose.
The Lectin-catch method using LecBeads (as shown in Application tab) is available for selective separation of glycoconjugates (glycoproteins, glycolipids, and so on). This rMOA1a-LecBeads (1 ml gel) shows a binding capacity of about 10 mg or more Cetuximab through its binding to a terminal [Galα(1-3)Gal] glycan epitope on Cetuximab.


  • Preparation : Recombinant MOA is covalently coupled with agarose gel
  • Quantity : 1 ml gel/2 ml (1:1, v/v slurry) suspended in PBS, pH7.5
  • Protein concentration : about 10 mg/ml gel
  • Binding capacity : >10 mg of glycoprotein (Cetuximab) /ml gel
  • Preservative : 0.02% [w/v] Sodium azide
  • Stabilizer : None
  • Store at 2-10 °C. (Avoid freezing).

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR