Isopropyl salicylate ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Isopropyl 2-hydroxybenzoate

S0014-25ML
TCIAS0014-25MLEA 515 NOK
TCIAS0014-25ML
Isopropyl salicylate ≥99.0% (by GC)
Isopropyl salicylate
Formel: C₁₀H₁₂O₃
MW: 180.2 g/mol
Kokepunkt: 235°C
MDL nummer: MFCD00035703
CAS-nummer: 607-85-2
EINECS: 210-143-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8818e563653aeed20a55126f9c046e1482d7b3dc&vid=8818e563653aeed20a55126f9c046e1482d7b3dc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR