Sodium pentacyanoammineferroate(II) hydrate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Trisodium Amminepentacyanoferroate(II), Pentacyanoammineferroate(II) Sodium Salt

S0050-25G
TCIAS0050-25GEA 1525 NOK
TCIAS0050-25G
Sodium pentacyanoammineferroate(II) hydrate ≥95.0% (by titrimetric analysis)
Sodium pentacyanoammineferroate(II) hydrate
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00059942
CAS-nummer: 14099-05-9
FN: 1588
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) min. 95.0 % (calcd.on anh.substance)
Solubility in Water transparency
Water 12.0 to 25.0 %
Sensitiveness Colorimetric Test : Turn Red purple with 10ppm Thiosemicarbazide


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR