Spermidine phosphate 98,0 Titrimetric

Leverandør: TCI
Warning

Synonymer: N-(3-Aminopropyl)-1,4-butanediamine Phosphate, N-(3-Aminopropyl)tetramethylenediamine Phosphate

S0068-1G S0068-5G
TCIAS0068-1GEA 460 NOK
TCIAS0068-1G TCIAS0068-5G
Spermidine phosphate 98,0 Titrimetric
Spermidine phosphate
Formel: C₇H₁₉N₃
MW: 439.23 g/mol
MDL nummer: MFCD00043283
CAS-nummer: 49721-50-8
EINECS: 256-447-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e583f49edc0dd892bbdaa84f3fa526af3f90838&vid=ad273632fe9804797ecc52ec4243dd8e4a63c922&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR