Dimethyl succinate ≥98.0% (by GC)

Leverandør: TCI
Warning

S0104-25G S0104-500G
TCIAS0104-25GEA 220 NOK
TCIAS0104-25G TCIAS0104-500G
Dimethyl succinate ≥98.0% (by GC)
Dimethyl succinate
Formel: C₆H₁₀O₄
Kokepunkt: 196°C
Smeltepunkt: 18…19 °C
Tetthet: 1,117 g/cm³ (20 °C)
Flash punkt: 98°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008466
CAS-nummer: 106-65-0
EINECS: 203-419-9
Merck Indeks: 12,09038
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60102bb8d52e3917d317c6f3a1c7846f9ab29850&vid=60102bb8d52e3917d317c6f3a1c7846f9ab29850&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR