Acid red 9

Leverandør: TCI

Synonymer: Silk Scarlet

S0140-25G
TCIAS0140-25GEA 475 NOK
TCIAS0140-25G
Acid red 9
Acid red 9
Formel: C₂₀H₁₄N₂O₄S
MDL nummer: MFCD00151319
CAS-nummer: 8003-59-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Lambda max. 492.0 to 496.0 nm (H2O)
Absorbance (E1%1cm) min. 400 (H2O, 492.0 to 496.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR