Safranine O

Leverandør: TCI
Warning

Synonymer: CI 50240

S0145-25G S0145-500G
TCIAS0145-25GEA 365 NOK
TCIAS0145-25G TCIAS0145-500G
Safranine O
Safranine O
Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011759
CAS-nummer: 477-73-6
EINECS: 207-518-8

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Lambda max. 517.0 to 522.0 nm (H2O)
Absorbance (E1%1cm) min. 550 (H2O, 517.0 to 522.0 nm) (calcd.on dried substance)
Drying loss max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR