Succinylcholine chloride dihydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Succinyldicholine dichloride, Suxamethonium chloride, Bis(trimethylammonioethyl) succinate chloride, Succinylcholine chloride

S0149-5G
TCIAS0149-5GEA 340 NOK
TCIAS0149-5G
Succinylcholine chloride dihydrate ≥98.0% (by titrimetric analysis)
Succinylcholine chloride dihydrate
Formel: C₁₄H₃₀Cl₂N₂O₄.2H₂O
MW: 397,34 g/mol
MDL nummer: MFCD00150564
CAS-nummer: 6101-15-1
EINECS: 200-747-4
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,08875

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Melting point 162.0 to 166.0 deg-C
Solubility in Water almost transparency
Water 8.0 to 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR