Scopolamine-N-oxide hydrobromide monohydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Genoscopolamine Hydrobromide, Scopolamine Aminoxide Hydrobromide

S0229-1G
TCIAS0229-1GEA 1150 NOK
TCIAS0229-1G
Scopolamine-N-oxide hydrobromide monohydrate ≥98.0% (by titrimetric analysis)
Scopolamine-N-oxide hydrobromide monohydrate
Formel: C₁₇H₂₁NO₅
MW: 400,27 g/mol
MDL nummer: MFCD00050590
CAS-nummer: 6106-81-6
EINECS: 228-066-8
FN: 1544
ADR: 6.1,III
Merck Indeks: 14,08407

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -22.0 to -25.0 deg (C=1,water)
Water 3.9 to 4.8 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR