Methyl hexanoate, standard material for GC

Leverandør: TCI

S0304-5ML
TCIAS0304-5MLEA 1200 NOK
TCIAS0304-5ML
Methyl hexanoate, standard material for GC
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71dc1f9f13bdae149b17e0eb92efe4ceb4e8f52f&vid=715b51f1d70b04e3cd2a9f679e50be0effc3ad85&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Methyl Hexanoate
utseende Colorless clear liquid
CAS-nr 106-70-7
Formel C₇H₁₄O₂
kokepunkt 150 °C (lit.)
Flammepunkt "46 °C (lit.)"

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR