SATA (N-Succinimidyl S-Acetylthioglycolate) ≥94.0% (by GC) cross-linking reagent

Leverandør: TCI
Warning

Synonymer: N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate

S0431-1G S0431-5G
TCIAS0431-1GEA 3100 NOK
TCIAS0431-1G TCIAS0431-5G
SATA (N-Succinimidyl S-Acetylthioglycolate) ≥94.0% (by GC) cross-linking reagent
SATA (N-Succinimidyl S-Acetylthioglycolate)
Formel: C₈H₉NO₅S
MW: 231.22 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036891
CAS-nummer: 76931-93-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 94.0 %
Melting point 95.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR