Standard mixture of fatty acid methyl esters (consists of methyl hexanoate, heptanoate, octanoate, nonanoate, and decanoate), standard material for GC
Leverandør: TCI
S0459-5ML
TCIAS0459-5MLEA 1550 NOK
TCIAS0459-5ML
Standard mixture of fatty acid methyl esters (consists of methyl hexanoate, heptanoate, octanoate, nonanoate, and decanoate), standard material for GC
Standarder PetroleumIndustristandarder