Spectinomycin dihydrochloride pentahydrate ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI

Synonymer: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

S0584-25G S0584-5G
TCIAS0584-25GEA 2400 NOK
TCIAS0584-25G TCIAS0584-5G
Spectinomycin dihydrochloride pentahydrate ≥98.0% (by total nitrogen and titration analysis)
Spectinomycin dihydrochloride pentahydrate
Formel: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Smeltepunkt: 207°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00150886
CAS-nummer: 22189-32-8
Merck Indeks: 14,08739

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +15.0 to +21.0 deg (C=1,H2O)
Water 16.0 to 20.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR