Sulphamonomethoxine ≥93.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 4-Amino-N-(6-methoxypyrimidin-4-yl)benzenesulphonamide, 4-Amino-N-(6-methoxy-4-pyrimidinyl)benzenesulphonamide, N-(6-Methoxy-4-pyrimidinyl)sulphanilamide

S0592-5G
TCIAS0592-5GEA 1175 NOK
TCIAS0592-5G
Sulphamonomethoxine ≥93.0% (by HPLC)
Sulphamonomethoxine
Formel: C₁₁H₁₂N₄O₃S
MW: 280,30 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00063466
CAS-nummer: 1220-83-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 201.0 to 206.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR